1,4-Diiodoperfluorobutane ≥98.0%

Supplier: TCI America

Synonyms: Octafluoro-1,4-diiodobutane, 1,4-Diiodooctafluorobutane

D2329-25G D2329-5G
TCD2329-25GEA 314.42 CAD
TCD2329-25G TCD2329-5G
1,4-Diiodoperfluorobutane ≥98.0%
1,4-Diiodoperfluorobutane

CAS Number: 375-50-8
MDL Number: MFCD00042263
Molecular Formula: C4F8I2
Molecular Weight: 453.84
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 63
Specific Gravity (20/20): 2.50


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Perfluoro-1,4-diiodobutane

Formula: C₄F₈I₂
MW: 453.84 g/mol
Boiling Pt: 150
Melting Pt: -3
Density: 2.50 (20 °C)
MDL Number: MFCD00042263
CAS Number: 375-50-8
EINECS: 206-788-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14cd29f9fc43f62d2649cae88f2f692c4c3800d1&vid=14cd29f9fc43f62d2649cae88f2f692c4c3800d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR