2,4-Dimethoxypyrimidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2327-25G D2327-5G
TCD2327-025GEA 173.8 CAD
TCD2327-025G 76219-004
2,4-Dimethoxypyrimidine ≥98.0% (by GC, titration analysis)
2,4-Dimethoxypyrimidine

CAS Number: 3551-55-1
MDL Number: MFCD00056028
Molecular Formula: C6H8N2O2
Molecular Weight: 140.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 92
Specific Gravity (20/20): 1.17

Formula: C₆H₈N₂O₂
Boiling Pt: 205 °C
Melting Pt: 17.5 °C
Density: 1.17 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00056028
CAS Number: 3551-55-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e4e60babb901d30fb7208eb3b27f1625797e351&vid=6e4e60babb901d30fb7208eb3b27f1625797e351&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR