1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22) ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D2323-5G D2323-1G
TCD2323-5GEA 944.84 CAD
TCD2323-5G TCD2323-001G
1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22) ≥98.0% (by titrimetric analysis)
1,10-Diaza-18-crown-6 (1,4,10,13-Tetraoxa-7,16-diazacyclooctadecane, Kryptofix® 22)

CAS Number: 23978-55-4
MDL Number: MFCD00005112
Molecular Formula: C12H26N2O4
Molecular Weight: 262.35
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 115


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 1,4,10,13-Tetraoxa-7,16-diazacyclooctadecane

Formula: C₁₂H₂₆N₂O₄
Melting Pt: 115 °C
MDL Number: MFCD00005112
CAS Number: 23978-55-4
EINECS: 245-965-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=413b7875125957b1ec236d1e257bb502ec775e3e&vid=413b7875125957b1ec236d1e257bb502ec775e3e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR