2,4-Dichloropyrimidine ≥98.0%

Supplier: TCI America
Warning

D2310-5G D2310-25G
TCD2310-005GEA 56.88 CAD
TCD2310-005G TCD2310-025G
2,4-Dichloropyrimidine ≥98.0%
2,4-Dichloropyrimidine

CAS Number: 3934-20-1
MDL Number: MFCD00006061
Molecular Formula: C4H2Cl2N2
Molecular Weight: 148.97
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 101
Melting point (°C): 60

Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 101 °C
Melting Pt: 60 °C
MDL Number: MFCD00006061
CAS Number: 3934-20-1
EINECS: 223-508-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19f72e9417fbe7aec4dcfe8b55985780b54b379f&vid=19f72e9417fbe7aec4dcfe8b55985780b54b379f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR