4,5-Dichloro-o-phenylenediamine 98,0

Supplier: TCI America

Synonyms: 4,5-Dichloro-1,2-phenylenediamine, 1,2-Diamino-4,5-dichlorobenzene, 4,5-Dichlorobenzene-1,2-diamine

D2307-25G D2307-5G
TCD2307-025GEA 426.6 CAD
TCD2307-025G TCD2307-005G
4,5-Dichloro-o-phenylenediamine 98,0
4,5-Dichloro-o-phenylenediamine

CAS Number: 5348-42-5
MDL Number: MFCD00007723
Molecular Formula: C6H6Cl2N2
Molecular Weight: 177.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 163
Flash Point (°C): 110


Other Product Names / Synonyms: 1,2-Diamino-4,5-dichlorobenzene

Formula: C₆H₆Cl₂N₂
MW: 177.03 g/mol
Boiling Pt: 305
Melting Pt: 163
Flash Pt: 110 °C
MDL Number: MFCD00007723
CAS Number: 5348-42-5
EINECS: 226-305-0
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37b0b65393d4a1548fad0ecfc3da7c0ac20d2a3f&vid=37b0b65393d4a1548fad0ecfc3da7c0ac20d2a3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR