2,3-Dichloroanisole ≥97.0%
Supplier: TCI America
D2306-500G
D2306-25G
TCD2306-500GEA
646.22
CAD
TCD2306-500G
TCD2306-025G
2,3-Dichloroanisole ≥97.0%
2,3-Dichloroanisole
CAS Number: 1984-59-4
MDL Number: MFCD00000545
Molecular Formula: C7H6Cl2O
Molecular Weight: 177.02
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Flash Point (°C): 113
Freezing point (°C): 30
Other Product Names / Synonyms: 2,3-Dichloromethoxybenzene, 1,2-Dichloro-3-methoxybenzene
Formula:
C₇H₆Cl₂O Boiling Pt: 128 °C (20 mmHg) Melting Pt: 31…34 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000545 CAS Number: 1984-59-4 EINECS: 217-853-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...