2,5-Dibromohexane ≥98.0%

Supplier: TCI America
Warning

D2298-5G D2298-25G
TCD2298-005GEA 86.9 CAD
TCD2298-005G TCD2298-025G
2,5-Dibromohexane ≥98.0%
2,5-Dibromohexane

CAS Number: 24774-58-1
MDL Number: MFCD00039177
Molecular Formula: C6H12Br2
Molecular Weight: 243.97
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.58

Formula: H₃CBrCH(CH₂)₂BrCHCH₃
Density: 1.58 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00039177
CAS Number: 24774-58-1
EINECS: 246-457-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebdcd2d5fc7164d483905a58cd04c69cf0072f7b&vid=ebdcd2d5fc7164d483905a58cd04c69cf0072f7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR