4,5-Dibromo-o-xylene ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1,2-Dibromo-4,5-dimethylbenzene, 4,5-Dibromo-1,2-dimethylbenzene
D2272-25G
D2272-5G
TCD2272-25GEA
649.54
CAD
TCD2272-25G
TCD2272-005G
4,5-Dibromo-o-xylene ≥98.0%
4,5-Dibromo-o-xylene
CAS Number: 24932-48-7
MDL Number: MFCD00082743
Molecular Formula: C8H8Br2
Molecular Weight: 263.96
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 278
Melting point (°C): 87
Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 4,5-Dibromo-o-xylene
Formula:
C₈H₈Br₂ MW: 263.96 g/mol Boiling Pt: 278 °C Melting Pt: 87 °C |
MDL Number:
MFCD00082743 CAS Number: 24932-48-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...