2,6-Dimethylbenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2260-25G D2260-5G
TCD2260-25GEA 328.64 CAD
TCD2260-25G TCD2260-005G
2,6-Dimethylbenzoic acid ≥98.0% (by GC, titration analysis)
2,6-Dimethylbenzoic acid

CAS Number: 632-46-2
MDL Number: MFCD00002483
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 160
Melting point (°C): 116


Other Product Names / Synonyms: m-Xylene-2-carboxylic Acid

Formula: C₉H₁₀O₂
Boiling Pt: 160 °C
Melting Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00002483
CAS Number: 632-46-2
EINECS: 211-177-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3e85804e9dac4c158d26b993206d9d5b5b6f7b4&vid=437a629fab2a6371c692167c7728bff2777ebfeb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR