2,6-Di-tert-butyl-p-benzoquinone ≥98.0%

Supplier: TCI America

Synonyms: 2,6-Di-tert-butylcyclohexa-2,5-diene-1,4-dione

D2256-25G D2256-5G
TCD2256-025GEA 371.3 CAD
TCD2256-025G TCD2256-005G
2,6-Di-tert-butyl-p-benzoquinone ≥98.0%
2,6-Di-tert-butyl-p-benzoquinone

CAS Number: 719-22-2
MDL Number: MFCD00001601
Molecular Formula: C14H20O2
Molecular Weight: 220.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 67


Other Product Names / Synonyms: 2,6-Di-tert-butyl-p-quinone

Formula: C₁₄H₂₀O₂
Melting Pt: 67 °C
MDL Number: MFCD00001601
CAS Number: 719-22-2
EINECS: 211-946-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ec0d8d56f8b603d884b277a62be1194337fd553&vid=4ec0d8d56f8b603d884b277a62be1194337fd553&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR