Diphenyliodonium hexafluorophosphate ≥97.0%

Supplier: TCI America

D2238-5G D2238-25G D2238-1G
TCD2238-5GEA 165.9 CAD
TCD2238-5G TCD2238-25G TCD2238-001G
Diphenyliodonium hexafluorophosphate ≥97.0%
Diphenyliodonium hexafluorophosphate

CAS Number: 58109-40-3
MDL Number: MFCD00061398
Molecular Formula: C12H10F6IP
Molecular Weight: 426.08
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 142

Formula: C₁₂H₁₀F₆IP
Melting Pt: 142 °C
MDL Number: MFCD00061398
CAS Number: 58109-40-3
EINECS: 261-134-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0025593916b98ef863cd4a206ef5c1ff74958a7a&vid=0025593916b98ef863cd4a206ef5c1ff74958a7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR