2,6-Dimethyl-p-benzoquinone ≥98.0%
Supplier: TCI America
D2234-5G
D2234-25G
D2234-1G
TCD2234-5GEA
494.54
CAD
TCD2234-5G
TCD2234-25G
TCD2234-001G
2,6-Dimethyl-p-benzoquinone ≥98.0%
2,6-Dimethyl-p-benzoquinone
CAS Number: 527-61-7
MDL Number: MFCD00001605
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 73
Product Categories: Synthetic Organic Chemistry, C-H Bond Activation Reaction, Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: 2,6-Dimethyl-2,5-cyclohexadiene-1,4-dione, 2,6-Dimethyl-p-quinone, m-Xyloquinone
Formula:
C₈H₈O₂ Melting Pt: 73 °C |
MDL Number:
MFCD00001605 CAS Number: 527-61-7 EINECS: 208-420-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...