2,3-Dimethylbenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D2221-25G D2221-5G
TCD2221-25GEA 114.08 CAD
TCD2221-25G TCD2221-005G
2,3-Dimethylbenzoic acid ≥98.0% (by titrimetric analysis)
2,3-Dimethylbenzoic acid

CAS Number: 603-79-2
MDL Number: MFCD00002479
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 145


Other Product Names / Synonyms: o-Xylene-3-carboxylic Acid

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 145 °C
Density: 1.283 g/cm³ (20 °C)
MDL Number: MFCD00002479
CAS Number: 603-79-2
EINECS: 210-058-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4cee24e5229a4358985595e479f3373ea973118&vid=9666d03e6e0345117e9e637de7ef52e67fd9df83&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR