Methyl-3,5-dihydroxybenzoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 3,5-dihydroxybenzoate, 3,5-Dihydroxybenzoic acid methyl ester

D2215-25G D2215-250G
TCD2215-025GEA 102.7 CAD
TCD2215-025G TCD2215-250G
Methyl-3,5-dihydroxybenzoate ≥98.0%
Methyl 3,5-dihydroxybenzoate

CAS Number: 2150-44-9
MDL Number: MFCD00002289
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 166


Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: 3,5-Dihydroxybenzoic Acid Methyl Ester, alpha-Resorcylic Acid Methyl Ester

Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 300
Melting Pt: 166 °C
MDL Number: MFCD00002289
CAS Number: 2150-44-9
EINECS: 218-426-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f9934c968802b5c972d6c9ce2b0339e486abf64&vid=9f9934c968802b5c972d6c9ce2b0339e486abf64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR