3,5-Dibromo-1-fluorobenzene ≥97.0%
Supplier: TCI America
Synonyms:
1,3-Dibromo-5-fluorobenzene
D2211-25G
D2211-10G
TCD2211-25GEA
139.04
CAD
TCD2211-25G
TCD2211-010G
3,5-Dibromo-1-fluorobenzene ≥97.0%
3,5-Dibromo-1-fluorobenzene
CAS Number: 1435-51-4
MDL Number: MFCD00061119
Molecular Formula: C6H3Br2F
Molecular Weight: 253.90
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 84
Flash Point (°C): 110
Specific Gravity (20/20): 2.03
Formula:
C₆H₃Br₂F MW: 253.9 g/mol Boiling Pt: 84 °C Density: 2.03 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00061119 CAS Number: 1435-51-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...