3,4-Dibutoxy-3-cyclobutene-1,2-dione ≥97.0%

Supplier: TCI America

Synonyms: Squaric acid dibutylester, Dibutyl squarate, 3,4-Dibutoxycyclobut-3-ene-1,2-dione, 1,2-Dibutoxy-1-cyclobutene-3,4-dione

D2203-5G D2203-25G
TCD2203-005GEA 109.02 CAD
TCD2203-005G TCD2203-025G
3,4-Dibutoxy-3-cyclobutene-1,2-dione ≥97.0%
3,4-Dibutoxy-3-cyclobutene-1,2-dione

CAS Number: 2892-62-8
MDL Number: MFCD00037150
Molecular Formula: C12H18O4
Molecular Weight: 226.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 113
Specific Gravity (20/20): 1.05


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: Dibutyl Squarate, Squaric Acid Dibutyl Ester, SADBE

Formula: C₁₂H₁₈O₄
MW: 226.27 g/mol
Boiling Pt: 121 °C (1013 hPa)
Density: 1.05 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00037150
CAS Number: 2892-62-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f00b16700b8d3cd437ce56c87fbe98775330175&vid=6f00b16700b8d3cd437ce56c87fbe98775330175&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR