3,6-Dithia-1,8-octanediol ≥98.0%
Supplier: TCI America
D2188-250G
D2188-25G
TCD2188-250GEA
377.62
CAD
TCD2188-250G
TCD2188-025G
3,6-Dithia-1,8-octanediol ≥98.0%
3,6-Dithia-1,8-octanediol
CAS Number: 5244-34-8
MDL Number: MFCD00002911
Molecular Formula: C6H14O2S2
Molecular Weight: 182.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 100
Melting point (°C): 65
Other Product Names / Synonyms: 1,2-Bis(2-hydroxyethylthio)ethane, 2,2'-(Ethylenedithio)diethanol
Formula:
C₆H₁₄O₂S₂ Boiling Pt: 100 °C Melting Pt: 65 °C |
MDL Number:
MFCD00002911 CAS Number: 5244-34-8 EINECS: 226-045-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...