trans-1,4-Diphenyl-2-butene-1,4-dione ≥98.0%
Supplier: TCI America
Synonyms:
1,2-Dibenzoylethylene, trans-1,2-Dibenzoylethylene
D2185-25G
D2185-5G
TCD2185-025GEA
509.08
CAD
TCD2185-025G
TCD2185-005G
trans-1,4-Diphenyl-2-butene-1,4-dione ≥98.0%
trans-1,4-Diphenyl-2-butene-1,4-dione
CAS Number: 959-28-4
MDL Number: MFCD00003083
Molecular Formula: C16H12O2
Molecular Weight: 236.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 110
Other Product Names / Synonyms: trans-Diphenacylidene, trans-1,4-Diphenyl-2-butene-1,4-dione
Formula:
C₁₆H₁₂O₂ Melting Pt: 110 °C |
MDL Number:
MFCD00003083 CAS Number: 959-28-4 EINECS: 213-498-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...