(1R,2R)-(+)-1,2-Diphenylethylenediamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2176-5G D2176-1G
TCD2176-5GEA 237.95 CAD
TCD2176-5G TCD2176-001G
(1R,2R)-(+)-1,2-Diphenylethylenediamine ≥98.0% (by GC, titration analysis)
(1R,2R)-(+)-1,2-Diphenylethylenediamine

CAS Number: 35132-20-8
MDL Number: MFCD00082769
Molecular Formula: C14H16N2
Molecular Weight: 212.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 83
Specific rotation [a]20/D: 103 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (1R,2R)-(+)-1,2-Diamino-1,2-diphenylethane

Formula: C₁₄H₁₆N₂
Melting Pt: 83 °C
MDL Number: MFCD00082769
CAS Number: 35132-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5caee00fcb76f4cca373b9f91896bf2c5bbe2cad&vid=5caee00fcb76f4cca373b9f91896bf2c5bbe2cad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR