(1S,2S)-(-)-1,2-Diphenylethylenediamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (1S,2S)-(-)-1,2-Diphenylethylenediamine, (1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine, (S,S)-DPEN

D2175-5G D2175-1G
TCD2175-5GEA 187.14 CAD
TCD2175-5G TCD2175-001G
(1S,2S)-(-)-1,2-Diphenylethylenediamine ≥98.0% (by GC, titration analysis)
(1S,2S)-(-)-1,2-Diphenylethylenediamine

CAS Number: 29841-69-8
MDL Number: MFCD00082751
Molecular Formula: C14H16N2
Molecular Weight: 212.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 83
Specific rotation [a]20/D: -103 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (1S,2S)-(-)-1,2-Diamino-1,2-diphenylethane

Formula: C₁₄H₁₆N₂
MW: 212.29 g/mol
Melting Pt: 83 °C
MDL Number: MFCD00082751
CAS Number: 29841-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b535d28386c3f217a40fad5db5a333898be97cd&vid=dacb825ae1f19c59019c1038091141eb4a01508e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR