1,8-Diiodooctane ≥95.0% stabilized
Supplier: TCI America
|
Warning
|
Stabilization:
Stabilised with Stabilised
D2158-5G
D2158-25G
TCD2158-5GEA
67.94
CAD
TCD2158-5G
TCD2158-025G
1,8-Diiodooctane ≥95.0% stabilized
1,8-Diiodooctane
(stabilized with Copper chip)
CAS Number: 24772-63-2
MDL Number: MFCD00001106
Molecular Formula: C8H16I2
Molecular Weight: 366.02
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 144
Flash Point (°C): 110
Specific Gravity (20/20): 1.84
Other Product Names / Synonyms: DIO
Formula:
C₈H₁₆I₂ Boiling Pt: 144 °C Melting Pt: 16…21 °C Density: 1.84 (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001106 CAS Number: 24772-63-2 EINECS: 246-456-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...