2,5-Dichlorotoluene ≥98.0%
Supplier: TCI America
Synonyms:
1,4-Dichloro-2-methylbenzene
D2157-5G
D2157-25G
TCD2157-5GEA
36.34
CAD
TCD2157-5G
TCD2157-025G
2,5-Dichlorotoluene ≥98.0%
2,5-Dichlorotoluene
CAS Number: 19398-61-9
MDL Number: MFCD00000609
Molecular Formula: C7H6Cl2
Molecular Weight: 161.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 199
Flash Point (°C): 111
Specific Gravity (20/20): 1.25
Formula:
C₇H₆Cl₂ Boiling Pt: 199 °C Density: 1.25 (20 °C) Flash Pt: 111 °C |
MDL Number:
MFCD00000609 CAS Number: 19398-61-9 EINECS: 243-032-2 UN: 3082 ADR: 9,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...