4,4'-Dimethoxybenzhydrol ≥98.0%
Supplier: TCI America
Synonyms:
4,4'-Dimethoxy-α-phenylbenzyl alcohol, Bis(4-methoxyphenyl)methanol, Bis(4-methoxyphenyl)carbinol, 4,4'-Dimethoxyphenyl phenyl carbinol, 4,4'-Dimethoxybenzhydryl alcohol, 4,4'-Dimethoxydiphenylmethanol
D2141-5G
D2141-25G
TCD2141-005GEA
75.84
CAD
TCD2141-005G
TCD2141-025G
4,4'-Dimethoxybenzhydrol ≥98.0%
4,4'-Dimethoxybenzhydrol
CAS Number: 728-87-0
MDL Number: MFCD00008410
Molecular Formula: C15H16O3
Molecular Weight: 244.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 70
Other Product Names / Synonyms: Bis(4-methoxyphenyl)methanol
Formula:
C₁₅H₁₆O₃ MW: 244.29 g/mol Boiling Pt: 362 Melting Pt: 70 °C |
MDL Number:
MFCD00008410 CAS Number: 728-87-0 EINECS: 211-975-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...