4,5-Diphenylimidazole 98,0

Supplier: TCI America

Synonyms: 4,5-Diphenyl-1H-imidazole

D2107-5G D2107-25G
TCD2107-005GEA 53.72 CAD
TCD2107-005G TCD2107-025G
4,5-Diphenylimidazole 98,0
4,5-Diphenylimidazole

CAS Number: 668-94-0
MDL Number: MFCD00005198
Molecular Formula: C15H12N2
Molecular Weight: 220.28
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 234

Formula: C₁₅H₁₂N₂
MW: 220.27 g/mol
Melting Pt: 234 °C
MDL Number: MFCD00005198
CAS Number: 668-94-0
EINECS: 211-573-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a05bbc9129fd2a8919c39934415c4a8fb961e42b&vid=a05bbc9129fd2a8919c39934415c4a8fb961e42b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR