3,4-Dimethoxy-3-cyclobutene-1,2-dione ≥98.0%

Supplier: TCI America

Synonyms: 1,2-Dimethoxy-1-cyclobutene-3,4-dione, Dimethyl squarate, Squaric acid dimethylester

D2099-25G D2099-5G D2099-1G
TCD2099-25GEA 726.8 CAD
TCD2099-25G TCD2099-5G TCD2099-001G
3,4-Dimethoxy-3-cyclobutene-1,2-dione ≥98.0%
3,4-Dimethoxy-3-cyclobutene-1,2-dione

CAS Number: 5222-73-1
MDL Number: MFCD00101316
Molecular Formula: C6H6O4
Molecular Weight: 142.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 56


Other Product Names / Synonyms: Dimethyl Squarate, Squaric Acid Dimethyl Ester

Formula: C₆H₆O₄
MW: 142.11 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 56 °C
MDL Number: MFCD00101316
CAS Number: 5222-73-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0e06182509627d3d7a46ec6504bed9559a8dda6&vid=e0e06182509627d3d7a46ec6504bed9559a8dda6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR