4,5-Dichloroimidazole ≥97.0%

Supplier: TCI America

Synonyms: 4,5-Dichloro-1H-imidazole

D2091-5G D2091-25G
TCD2091-5GEA 134.3 CAD
TCD2091-5G TCD2091-025G
4,5-Dichloroimidazole ≥97.0%
4,5-Dichloroimidazole

CAS Number: 15965-30-7
MDL Number: MFCD00005195
Molecular Formula: C3H2Cl2N2
Molecular Weight: 136.96
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 181

Formula: C₃H₂Cl₂N₂
Melting Pt: 181 °C
MDL Number: MFCD00005195
CAS Number: 15965-30-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49e67578f8ed380c334b37cbb134971f89f345ca&vid=49e67578f8ed380c334b37cbb134971f89f345ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR