2,3-Difluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D2058-25G D2058-5G
TCD2058-25GEA 459.46 CAD
TCD2058-25G TCD2058-005G
2,3-Difluorobenzoic acid ≥98.0% (by GC, titration analysis)
2,3-Difluorobenzoic acid

CAS Number: 4519-39-5
MDL Number: MFCD00010267
Molecular Formula: C7H4F2O2
Molecular Weight: 158.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 164

Formula: C₇H₄F₂O₂
Melting Pt: 164 °C
MDL Number: MFCD00010267
CAS Number: 4519-39-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b2252a0f858f8b66abea7cc6808ba2fd24709a3&vid=ac1e66d5eca2cbeb15c2d449205fa6db08624651&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR