1,4-Diphenoxybenzene ≥98.0%

Supplier: TCI America

D2017-25G D2017-5G
TCD2017-25GEA 270.18 CAD
TCD2017-25G TCD2017-005G
1,4-Diphenoxybenzene ≥98.0%
1,4-Diphenoxybenzene

CAS Number: 3061-36-7
MDL Number: MFCD00038368
Molecular Formula: C18H14O2
Molecular Weight: 262.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 182
Melting point (°C): 76


Product Categories: Materials Chemistry, Diphenyl Ethers (for High-Performance Polymer Research), Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Hydroquinone Diphenyl Ether

Formula: C₁₈H₁₄O₂
Boiling Pt: 182 °C
Melting Pt: 76 °C
MDL Number: MFCD00038368
CAS Number: 3061-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13a49008659f0347def1f7b84f2b52c989a3c4d2&vid=13a49008659f0347def1f7b84f2b52c989a3c4d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR