Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Dimethylpropyleneurea, Dimethyl propylene urea, 1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 1,3-Dimethyltetrahydropyrimidin-2(1H)-one, DMPU, 1,3-Dimethyltetrahydro-2(1H)-pyrimidinone, N,N'-Dimethyl propylene urea
CAS Number: 7226-23-5
MDL Number: MFCD00006550
Molecular Formula: C6H12N2O
Molecular Weight: 128.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): 121
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: N,N'-Dimethylpropyleneurea, DMPU
Formula:
C₆H₁₂N₂O MW: 128.17 g/mol Boiling Pt: 93 °C Melting Pt: –24 °C Density: 1.07 (20 °C) Flash Pt: 121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006550 CAS Number: 7226-23-5 EINECS: 230-625-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...