4,4'-Diaminobenzanilide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
4-Amino-N-(4-aminophenyl)benzamide
D2005-25G
D2005-500G
TCD2005-025GEA
105.86
CAD
TCD2005-025G
TCD2005-500G
4,4'-Diaminobenzanilide ≥98.0% (by HPLC, titration analysis)
4,4'-Diaminobenzanilide
CAS Number: 785-30-8
MDL Number: MFCD00025361
Molecular Formula: C13H13N3O
Molecular Weight: 227.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Flash Point (°C): 100
Formula:
C₁₃H₁₃N₃O MW: 227.27 g/mol Melting Pt: 214…215 °C Flash Pt: 100 °C |
MDL Number:
MFCD00025361 CAS Number: 785-30-8 EINECS: 212-321-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...