Tris(triphenylphosphine)ruthenium(II) chloride ≥90.0%

Supplier: TCI America

Synonyms: Dichlorotris(triphenylphosphine)ruthenium(II), Dichlorotris(triphenylphosphino)ruthenium (II)

D1997-1G D1997-5G
TCD1997-001GEA 83.74 CAD
TCD1997-001G TCD1997-005G
Tris(triphenylphosphine)ruthenium(II) chloride ≥90.0%
Tris(triphenylphosphine)ruthenium (II) chloride

CAS Number: 15529-49-4
MDL Number: MFCD00013077
Molecular Formula: C54H45Cl2P3Ru
Molecular Weight: 958.85
Purity/Analysis Method: >90.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, C-H Bond Activation Reaction, Oxidation, Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: Dichlorotris(triphenylphosphine)ruthenium(II)

Formula: C₅₄H₄₅Cl₂P₃Ru
MW: 958.85 g/mol
MDL Number: MFCD00013077
CAS Number: 15529-49-4
EINECS: 239-569-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdd28870f8c518746272f797c710e60990e1adf1&vid=bdd28870f8c518746272f797c710e60990e1adf1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR