2,5-Dibromoaniline ≥98.0%
Supplier: TCI America
D1989-5G
D1989-25G
TCD1989-005GEA
64.78
CAD
TCD1989-005G
TCD1989-025G
2,5-Dibromoaniline ≥98.0%
2,5-Dibromoaniline
CAS Number: 3638-73-1
MDL Number: MFCD00007636
Molecular Formula: C6H5Br2N
Molecular Weight: 250.92
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 56
Flash Point (°C): 109
Formula:
C₆H₅Br₂N Boiling Pt: 281 °C (1013 hPa) Melting Pt: 56 °C Flash Pt: 109 °C |
MDL Number:
MFCD00007636 CAS Number: 3638-73-1 EINECS: 222-865-5 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...