4-Methylveratrole ≥99.0%

Supplier: TCI America
Warning

Synonyms: 1,2-Dimethoxy-4-methylbenzene, 3,4-Dimethoxytoluene

D1986-25G D1986-500G D1986-100G
TCD1986-025GEA 26.86 CAD
TCD1986-025G TCD1986-500G TCD1986-100G
4-Methylveratrole ≥99.0%
4-Methylveratrole

CAS Number: 494-99-5
MDL Number: MFCD00016651
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Flash Point (°C): 85
Freezing point (°C): 22


Other Product Names / Synonyms: Homoveratrole

Formula: C₉H₁₂O₂
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00016651
CAS Number: 494-99-5
EINECS: 207-796-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c650d59d5da106de828fd131427b8433f131debc&vid=d96dafe4b2e3b4890d28d842a3527e50833a3fc3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR