1,2-Epoxydodecane ≥95.0%
Supplier: TCI America
D1984-25ML
D1984-100ML
TCD1984-025MLEA
49.82
CAD
TCD1984-025ML
TCD1984-100ML
1,2-Epoxydodecane ≥95.0%
1,2-Epoxydodecane
CAS Number: 2855-19-8
MDL Number: MFCD00005150
Molecular Formula: C12H24O
Molecular Weight: 184.32
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 105
Specific Gravity (20/20): 0.84
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1-Dodecene Epoxide, 1,2-Dodecylene Oxide
Formula:
C₁₂H₂₄O Boiling Pt: 85 °C Density: 0.84 (20 °C) Flash Pt: 105 °C |
MDL Number:
MFCD00005150 CAS Number: 2855-19-8 EINECS: 220-667-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...