3',5'-Dihydroxyacetophenone ≥98.0%

Supplier: TCI America

Synonyms: 1-(3,5-Dihydroxyphenyl)ethan-1-one, 1-(3,5-Dihydroxyphenyl)ethanone

D1981-25G D1981-10G
TCD1981-025GEA 197.5 CAD
TCD1981-025G TCD1981-010G
3',5'-Dihydroxyacetophenone ≥98.0%
3',5'-Dihydroxyacetophenone

CAS Number: 51863-60-6
MDL Number: MFCD00002290
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 147


Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: 5-Acetylresorcinol

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 147 °C
MDL Number: MFCD00002290
CAS Number: 51863-60-6
EINECS: 257-480-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d31a7bf01a38abdf04c8f0828fa79fbbcbfc7302&vid=d31a7bf01a38abdf04c8f0828fa79fbbcbfc7302&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR