4,4'-Dibromobenzil ≥97.0%

Supplier: TCI America
Warning

D1979-25G D1979-5G
TCD1979-25GEA 499.12 CAD
TCD1979-25G TCD1979-005G
4,4'-Dibromobenzil ≥97.0%
4,4'-Dibromobenzil

CAS Number: 35578-47-3
MDL Number: MFCD00000104
Molecular Formula: C14H8Br2O2
Molecular Weight: 368.02
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 227

Formula: C₁₄H₈Br₂O₂
Melting Pt: 227 °C
MDL Number: MFCD00000104
CAS Number: 35578-47-3
EINECS: 252-628-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=972af1269f49a8b1de2123969ac9fcb2107c5b57&vid=972af1269f49a8b1de2123969ac9fcb2107c5b57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR