Didodecyldimethylammonium bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D1974-500G D1974-25G D1974-100G
TCD1974-500GEA 1318.98 CAD
TCD1974-500G TCD1974-025G TCD1974-100G
Didodecyldimethylammonium bromide ≥98.0% (by titrimetric analysis)
Didodecyldimethylammonium bromide

CAS Number: 3282-73-3
MDL Number: MFCD00041969
Molecular Formula: C26H56BrN
Molecular Weight: 462.65
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 161


Other Product Names / Synonyms: Didodecyldimethylammonium Bromide

Formula: C₂₆H₅₆BrN
Melting Pt: 161 °C
MDL Number: MFCD00041969
CAS Number: 3282-73-3
EINECS: 221-923-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=068ff02ccb90f92fadcb1b73b51bf7f22d2334aa&vid=068ff02ccb90f92fadcb1b73b51bf7f22d2334aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR