2,3-Dimethoxy-5-methyl-p-benzoquinone ≥97.0%

Supplier: TCI America

Synonyms: 2,3-Dimethoxy-5-methylcyclohexa-2,5-diene-1,4-dione, 2,3-Dimethoxy-5-methyl-1,4-benzoquinone

D1956-5G D1956-1G
TCD1956-5GEA 205.4 CAD
TCD1956-5G TCD1956-001G
2,3-Dimethoxy-5-methyl-p-benzoquinone ≥97.0%
2,3-Dimethoxy-5-methyl-p-benzoquinone

CAS Number: 605-94-7
MDL Number: MFCD00001595
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 60


Product Categories: Materials Chemistry, Molecular Conductors, Bioscience, Vitamin Related Compounds
Other Product Names / Synonyms: Coenzyme Qo

Formula: C₉H₁₀O₄
Melting Pt: 60 °C
MDL Number: MFCD00001595
CAS Number: 605-94-7
EINECS: 210-100-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3bc6eff6cfab442b3fdb95c04a95d2f37d34e99&vid=e3bc6eff6cfab442b3fdb95c04a95d2f37d34e99&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR