2',5'-Dihydroxyacetophenone ≥98.0%

Supplier: TCI America

Synonyms: 1-(2,5-Dihydroxyphenyl)ethan-1-one, 1-(2,5-Dihydroxyphenyl)ethanone, 2,5-Dihydroxyacetophenone

D1955-25G D1955-5G
TCD1955-025GEA 297.04 CAD
TCD1955-025G TCD1955-005G
2',5'-Dihydroxyacetophenone ≥98.0%
2',5'-Dihydroxyacetophenone

CAS Number: 490-78-8
MDL Number: MFCD00002343
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 207


Other Product Names / Synonyms: 2-Acetylhydroquinone, Quinacetophenone

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 207 °C
MDL Number: MFCD00002343
CAS Number: 490-78-8
EINECS: 207-716-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ced0cd31753b8f35f42549d2812b024ddf8e3b8&vid=1ced0cd31753b8f35f42549d2812b024ddf8e3b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR