3,4-Diaminobenzophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D1950-25G D1950-100G
TCD1950-025GEA 343.81 CAD
TCD1950-025G TCD1950-100G
3,4-Diaminobenzophenone ≥98.0% (by GC, titration analysis)
3,4-Diaminobenzophenone

CAS Number: 39070-63-8
MDL Number: MFCD00007727
Molecular Formula: C13H12N2O
Molecular Weight: 212.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 118

Formula: C₁₃H₁₂N₂O
Boiling Pt: 390 °C (1013 hPa)
Melting Pt: 118 °C
MDL Number: MFCD00007727
CAS Number: 39070-63-8
EINECS: 254-273-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94a49703732678a2e1393a5211201db081f6a88e&vid=94a49703732678a2e1393a5211201db081f6a88e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR