2,4-Dichlorothiophenol ≥97.0%

Supplier: TCI America
Warning

Synonyms: 2,4-Dichlorobenzenethiol, 1,3-Dichloro-4-mercaptobenzene, 2,4-Dichlorobenzene-1-thiol

D1938-250G D1938-25G
TCD1938-250GEA 307.47 CAD
TCD1938-250G TCD1938-25G
2,4-Dichlorothiophenol ≥97.0%
2,4-Dichlorothiophenol

CAS Number: 1122-41-4
MDL Number: MFCD00041421
Molecular Formula: C6H4Cl2S
Molecular Weight: 179.06
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Freezing point (°C): 15
Specific Gravity (20/20): 1.42


Other Product Names / Synonyms: 2,4-Dichlorothiophenol

Formula: C₆H₄Cl₂S
MW: 179.07 g/mol
Boiling Pt: 115 °C
Melting Pt: 22 °C
Density: 1.42 (20 °C)
MDL Number: MFCD00041421
CAS Number: 1122-41-4
EINECS: 214-346-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83d63cdfef00ecd60905272234511bfb04587397&vid=0d3a2dcad43bc9c33fbd50ac0f256c38a459d683&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR