trans,trans-2,4-Decadienal ≥90.0%
Supplier: TCI America
D1934-5ML
D1934-25ML
TCD1934-005MLEA
66.04
CAD
TCD1934-005ML
TCD1934-25ML
trans,trans-2,4-Decadienal ≥90.0%
trans,trans-2,4-Decadienal
CAS Number: 25152-84-5
MDL Number: MFCD00007007
Molecular Formula: C10H16O
Molecular Weight: 152.24
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 100
Specific Gravity (20/20): 0.87
Formula:
C₁₀H₁₆O Boiling Pt: 116 °C Density: 0.87 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00007007 CAS Number: 25152-84-5 EINECS: 246-668-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...