1,2-Difluoro-4-nitrobenzene ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
3,4-Difluoronitrobenzene
D1926-250G
D1926-25G
TCD1926-250GEA
486.64
CAD
TCD1926-250G
TCD1926-025G
1,2-Difluoro-4-nitrobenzene ≥98.0%
1,2-Difluoro-4-nitrobenzene
CAS Number: 369-34-6
MDL Number: MFCD00007198
Molecular Formula: C6H3F2NO2
Molecular Weight: 159.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 78
Melting point (°C): -12
Flash Point (°C): 80
Specific Gravity (20/20): 1.45
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Nitro-3,4-difluorobenzene
Formula:
C₆H₃F₂NO₂ Boiling Pt: 78 °C Melting Pt: –12 °C Density: 1.45 (20 °C) Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007198 CAS Number: 369-34-6 EINECS: 206-718-2 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...