2,6-Diphenylpyridine ≥99.0%
Supplier: TCI America
|
Warning
|
D1922-25G
D1922-5G
TCD1922-25GEA
485.06
CAD
TCD1922-25G
TCD1922-005G
2,6-Diphenylpyridine ≥99.0%
2,6-Diphenylpyridine
CAS Number: 3558-69-8
MDL Number: MFCD00006284
Molecular Formula: C17H13N
Molecular Weight: 231.30
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 398
Formula:
C₁₇H₁₃N Boiling Pt: 398 °C Melting Pt: 85 °C |
MDL Number:
MFCD00006284 CAS Number: 3558-69-8 EINECS: 222-620-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...