4,4'-Dichlorobenzhydrol ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-Dichlorobenzhydryl alcohol, DBH, 4,4'-Dichlorophenyl phenyl carbinol, 4,4'-Dichloro-α-phenylbenzyl alcohol, 4,4'-Dichlorodiphenylmethanol, Bis(4-chlorophenyl)methanol, Bis(4-chlorophenyl)carbinol

D1915-250G D1915-25G
TCD1915-250GEA 685.72 CAD
TCD1915-250G TCD1915-025G
4,4'-Dichlorobenzhydrol ≥98.0%
4,4'-Dichlorobenzhydrol

CAS Number: 90-97-1
MDL Number: MFCD00000629
Molecular Formula: C13H10Cl2O
Molecular Weight: 253.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 93

Formula: C₁₃H₁₀Cl₂O
MW: 253.13 g/mol
Melting Pt: 93 °C
MDL Number: MFCD00000629
CAS Number: 90-97-1
EINECS: 202-029-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ca624424b9daea1e5f3a5eca013a51b7167e32f&vid=9ca624424b9daea1e5f3a5eca013a51b7167e32f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR