3,4-Difluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D1908-25G D1908-5G
TCD1908-25GEA 368.14 CAD
TCD1908-25G TCD1908-5G
3,4-Difluorobenzoic acid ≥98.0% (by GC, titration analysis)
3,4-Difluorobenzoic acid

CAS Number: 455-86-7
MDL Number: MFCD00011672
Molecular Formula: C7H4F2O2
Molecular Weight: 158.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 123

Formula: C₇H₄F₂O₂
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 123 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011672
CAS Number: 455-86-7
EINECS: 207-249-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=061677188766581c173d1173ab90b75a40651605&vid=12675f774b0745bd1ebe96492061e108591a23d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR