3,5-Dichlorothiophenol ≥97.0%

Supplier: TCI America
Warning

Synonyms: 3,5-Dichlorobenzene-1-thiol, 3,5-Dichlorobenzenethiol, 1,3-Dichloro-5-mercaptobenzene

D1905-5G D1905-25G
TCD1905-005GEA 127.35 CAD
TCD1905-005G TCD1905-025G
3,5-Dichlorothiophenol ≥97.0%
3,5-Dichlorothiophenol

CAS Number: 17231-94-6
MDL Number: MFCD00041423
Molecular Formula: C6H4Cl2S
Molecular Weight: 179.06
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 114
Melting point (°C): 63


Other Product Names / Synonyms: 3,5-Dichlorothiophenol

Formula: C₆H₄Cl₂S
MW: 179.07 g/mol
Boiling Pt: 114 °C
Melting Pt: 63 °C
MDL Number: MFCD00041423
CAS Number: 17231-94-6
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c5659dcca237bc98e14637a15aaf1b6e94f8f1f&vid=fa040c4c475eacec1aa736bb252c4c67e03a32a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR