2,6-Dichlorothiophenol ≥95.0%
Supplier: TCI America
|
Warning
|
D1904-5G
D1904-25G
TCD1904-005GEA
81.05
CAD
TCD1904-005G
TCD1904-025G
2,6-Dichlorothiophenol ≥95.0%
2,6-Dichlorothiophenol
CAS Number: 24966-39-0
MDL Number: MFCD00004833
Molecular Formula: C6H4Cl2S
Molecular Weight: 179.06
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 47
Flash Point (°C): 112
Other Product Names / Synonyms: 2,6-Dichlorothiophenol
Formula:
C₆H₄Cl₂S Boiling Pt: 246 °C (1013 hPa) Melting Pt: 47 °C Flash Pt: 112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004833 CAS Number: 24966-39-0 EINECS: 416-720-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...