2,3-Dichlorothiophenol ≥95.0%

Supplier: TCI America

D1903-25G D1903-5G
TCD1903-25GEA 357.08 CAD
TCD1903-25G TCD1903-005G
2,3-Dichlorothiophenol ≥95.0%
2,3-Dichlorothiophenol

CAS Number: 17231-95-7
MDL Number: MFCD00041420
Molecular Formula: C6H4Cl2S
Molecular Weight: 179.06
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 100
Melting point (°C): 59
Flash Point (°C): 110


Other Product Names / Synonyms: 2,3-Dichlorothiophenol

Formula: C₆H₄Cl₂S
Boiling Pt: 100 °C
Melting Pt: 59 °C
Flash Pt: 110 °C
MDL Number: MFCD00041420
CAS Number: 17231-95-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb0bf8b4a59bd6032ff73980ff864628bde37f00&vid=40383f62e8d0c7cb21ac4e18d1fbc0d2cfa3662f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR