9-Decen-1-ol ≥97.0%

Supplier: TCI America

D1892-25ML D1892-500ML
TCD1892-025MLEA 91.64 CAD
TCD1892-025ML TCD1892-500ML
9-Decen-1-ol ≥97.0%
9-Decen-1-ol

CAS Number: 13019-22-2
MDL Number: MFCD00002992
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 114
Specific Gravity (20/20): 0.85

Formula: C₁₀H₂₀O
Boiling Pt: 108 °C
Melting Pt: –13 °C
Density: 0.85 (20 °C)
Flash Pt: 114 °C
MDL Number: MFCD00002992
CAS Number: 13019-22-2
EINECS: 235-878-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5745792200cc2d47883fd990b45732732ca4ee6a&vid=5745792200cc2d47883fd990b45732732ca4ee6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR